This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03376081_u1
See other ACTG1 GE Assays ›
Gene Symbol:
ACTG1
Gene Name:
actin gamma 1
Gene Aliases:
actin
Chromosome Location:
Chr.12: 1320355 - 1323219 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03376081_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACTG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003357928.4 XP_003357976.1 1513 77
GenBank mRNA AK237086.1 - 5 1456 77
AK240355.1 - 3 918 77
AK392362.1 - 5 1459 77
AK395177.1 - 1712 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Endogenous Controls Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0