This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03374719_u1
See other PRKACA GE Assays ›
Gene Symbol:
PRKACA
Gene Name:
protein kinase cAMP-activated catalytic subunit alpha
Gene Aliases:
PKA
Chromosome Location:
Chr.2: 65074876 - 65095564 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03374719_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRKACA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003123353.4 XP_003123401.1 1046 77
XM_003354103.4 XP_003354151.1 821 77
GenBank mRNA X07617.1 - 3 248 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase A signaling
cellular response to heat
cellular response to glucose stimulus
negative regulation of meiotic cell cycle process involved in oocyte maturation
protein serine/threonine kinase activity
AMP-activated protein kinase activity
cAMP-dependent protein kinase activity
ATP binding
protein kinase A regulatory subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0