This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03374343_g1
See other UBC GE Assays ›
Gene Symbol:
UBC
Gene Name:
ubiquitin C
Gene Aliases:
-
Chromosome Location:
Chr.14: 28253857 - 28260771 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03374343_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003483411.4 XP_003483459.2 3094 85
XM_021074017.1 XP_020929676.1 3043 85
XM_021074018.1 XP_020929677.1 2950 85
GenBank mRNA AK346438.1 - 3 1201 85
AK348465.1 - 3 1201 85
AK391318.1 - 5 1658 85
AK395186.1 - 4 1215 85
AK400291.1 - 3 1201 85
M18159.1 - 1 757 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0