This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03374228_m1
See other GSK3B GE Assays ›
Gene Symbol:
GSK3B
Gene Name:
glycogen synthase kinase 3 beta
Gene Aliases:
GSK3beta
Chromosome Location:
Chr.13: 140136109 - 140370197 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03374228_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSK3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128443.1 NP_001121915.1 4 - 5 502 80
XM_021068182.1 XP_020923841.1 1559 80
XM_021068185.1 XP_020923844.1 1559 80
XM_021068186.1 XP_020923845.1 5659 80
XM_021068187.1 XP_020923846.1 1559 80
GenBank mRNA AK347804.1 - 4 - 5 847 80
EU586114.1 - 4 - 5 502 80
JN387127.1 - 4 - 5 596 80
JN387128.1 - 4 - 5 466 80
JN387129.1 - 4 - 5 466 80
JN387130.1 - 4 - 5 466 80
JN387131.1 - 4 - 5 466 80
JN387132.1 - 4 - 5 466 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0