This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss00943854_m1
See other UBE2V2 GE Assays ›
Gene Symbol:
UBE2V2
Gene Name:
ubiquitin conjugating enzyme E2 V2
Gene Aliases:
-
Chromosome Location:
Chr.4: 79621343 - 79652502 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss00943854_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2V2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243391.1 NP_001230320.1 1 - 2 78 77
GenBank mRNA AK231985.1 - 1 - 2 66 77
AK352057.1 - 1 - 2 78 77
AK395811.1 - 1 - 2 89 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001243391.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0