This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04109589_s1
See other CDC34 GE Assays ›
Gene Symbol:
CDC34
Gene Name:
Gene Aliases:
DNA6, UBC3, YDR054C
Chromosome Location:
Chr.IV: 561440 - 562327 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04109589_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDC34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001180362.1 NP_010339.1 1 555 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
protein polyubiquitination
DNA replication
ubiquitin-dependent protein catabolic process
cell cycle
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
protein autoubiquitination
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
transferase activity
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0