This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07365889_s1
See other LOC685619 GE Assays ›
Gene Symbol:
LOC685619
Gene Name:
similar to TP53-regulating kinase (p53-related protein kinase) (Nori-2)
Gene Aliases:
-
Chromosome Location:
Chr.3: 162164354 - 162168263 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07365889_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC685619
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002729250.4 XP_002729296.1 1 713 58
XM_006235633.2 XP_006235695.1 1 710 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_002729250.4, XM_006235633.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
tRNA threonylcarbamoyladenosine metabolic process
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header