This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07316474_m1
See other AHSA2 GE Assays ›
Gene Symbol:
Ahsa2
Gene Name:
AHA1, activator of heat shock protein ATPase 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 108262795 - 108273989 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07316474_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ahsa2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107241.1 NP_001100711.1 6 - 7 767 71
XM_006251539.2 XP_006251601.1 7 - 8 902 71
XM_006251540.2 XP_006251602.1 6 - 7 790 71
XM_006251541.2 XP_006251603.1 6 - 7 784 71
XM_006251542.2 XP_006251604.1 6 - 7 817 71
GenBank mRNA BC099114.1 - 7 - 8 806 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0