This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07314762_m1
See other FFAR4 GE Assays ›
Gene Symbol:
Ffar4
Gene Name:
free fatty acid receptor 4
Gene Aliases:
Gpr120, O3far1
Chromosome Location:
Chr.1: 256786124 - 256804156 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07314762_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ffar4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001047088.1 NP_001040553.1 1 - 2 610 92
GenBank mRNA AB207868.1 - 1 - 2 610 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
regulation of glucose transport
cellular response to hormone stimulus
negative regulation of apoptotic process
hormone secretion
negative regulation of cytokine secretion
negative regulation of inflammatory response
detection of chemical stimulus involved in sensory perception of taste
positive regulation of ERK1 and ERK2 cascade
response to peptide
G-protein coupled receptor activity
fatty acid binding
taste receptor activity
peptide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0