This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07314694_s1
See other RPRM GE Assays ›
Gene Symbol:
Rprm
Gene Name:
reprimo, TP53 dependent G2 arrest mediator candidate
Gene Aliases:
-
Chromosome Location:
Chr.3: 39595297 - 39598371 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07314694_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rprm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044276.1 NP_001037741.1 1 220 101
XM_008761775.1 XP_008759997.1 2 1769 101
GenBank mRNA BC091400.1 - 1 220 7323940 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC091400.1, NM_001044276.1, XM_008761775.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cell cycle arrest
regulation of mitotic cell cycle

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0