This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07314355_m1
See other MAGEB16 GE Assays ›
Gene Symbol:
Mageb16
Gene Name:
melanoma antigen family B, 16
Gene Aliases:
Snca
Chromosome Location:
Chr.X: 46977655 - 47026377 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07314355_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mageb16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025013.1 NP_001020184.1 2 - 3 144 101
XM_006256976.1 XP_006257038.1 3 - 4 204 101
XM_006256977.2 XP_006257039.1 4 - 5 267 101
GenBank mRNA BC079208.1 - 2 - 3 144 7131795 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC079208.1, NM_001025013.1, XM_006256976.1, XM_006256977.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mageb16

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0