This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07314213_m1
See other UBE2V2 GE Assays ›
Gene Symbol:
Ube2v2
Gene Name:
ubiquitin-conjugating enzyme E2 variant 2
Gene Aliases:
MMS2
Chromosome Location:
Chr.11: 89557273 - 89591175 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07314213_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2v2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183052.2 NP_898875.1 1 - 2 55 81
GenBank mRNA FM082155.1 - 1 - 2 45 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_183052.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2v2

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

DNA double-strand break processing
postreplication repair
positive regulation of neuron projection development
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
error-free postreplication DNA repair
negative regulation of neuron apoptotic process
positive regulation of DNA repair
positive regulation of synapse assembly
protein K63-linked ubiquitination
molecular_function
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0