This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07313512_g1
See other CALCB GE Assays ›
Gene Symbol:
Calcb
Gene Name:
calcitonin-related polypeptide, beta
Gene Aliases:
-
Chromosome Location:
Chr.1: 184271033 - 184276480 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07313512_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Calcb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138513.1 NP_612522.1 3 - 4 435 126
XM_006230075.2 XP_006230137.1 5 - 6 909 125
XM_008759674.1 XP_008757896.1 6 - 7 764 125
GenBank mRNA M11596.1 - 3 - 4 435 126

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Calcb

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Process(es)

hormone activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0