This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07313024_g1
See other NPPC GE Assays ›
Gene Symbol:
Nppc
Gene Name:
natriuretic peptide C
Gene Aliases:
-
Chromosome Location:
Chr.9: 93731436 - 93735636 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07313024_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nppc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053750.1 NP_446202.1 1 - 2 206 93
GenBank mRNA D90219.1 - 1 - 2 206 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nppc

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

ossification
response to hypoxia
growth plate cartilage chondrocyte differentiation
growth plate cartilage chondrocyte proliferation
cGMP biosynthetic process
receptor guanylyl cyclase signaling pathway
negative regulation of cell proliferation
post-embryonic development
regulation of cAMP metabolic process
regulation of cGMP metabolic process
positive regulation of cGMP biosynthetic process
regulation of multicellular organism growth
response to drug
single organism reproductive process
response to ethanol
positive regulation of osteoblast differentiation
positive regulation of vasodilation
animal organ development
regulation of smooth muscle cell proliferation
regulation of blood vessel size
negative regulation of DNA metabolic process
negative regulation of meiotic cell cycle
negative regulation of oocyte maturation
receptor binding
hormone activity
protein homodimerization activity
peptide hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0