This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07310147_m1
See other S100A13 GE Assays ›
Gene Symbol:
S100a13
Gene Name:
S100 calcium binding protein A13
Gene Aliases:
S100A1
Chromosome Location:
Chr.2: 189906022 - 189912516 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07310147_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191607.1 NP_001178536.1 2 - 3 248 93
GenBank mRNA AW141940.1 - 2 - 3 242 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell proliferation
regulation of cell shape
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to copper ion
cytokine secretion
interleukin-1 alpha secretion
response to electrical stimulus
molecular_function
copper ion binding
calcium ion binding
zinc ion binding
fibroblast growth factor binding
protein homodimerization activity
RAGE receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0