This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06432853_s1
See other LOC102554620 GE Assays ›
Gene Symbol:
LOC102554620
Gene Name:
von Willebrand factor C and EGF domain-containing protein-like
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_007906915: 65166 - 86730 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06432853_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102554620
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008773895.1 XP_008772117.1 1985 88
XM_008773897.1 XP_008772119.1 2476 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header