This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06430517_s1
See other LOC683722 GE Assays ›
Gene Symbol:
LOC683722
Gene Name:
Fgfr1 oncogene partner
Gene Aliases:
Fgfr1op
Chromosome Location:
Chr.1: 53122330 - 53153108 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06430517_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC683722
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101008.1 NP_001094478.1 1 137 65
XM_008758772.1 XP_008756994.1 1 389 65
XM_008758773.1 XP_008756995.1 1 386 65
XM_008758774.1 XP_008756996.1 1 242 65
GenBank mRNA BC098026.1 - 1 137 7455942 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
positive regulation of cell proliferation
positive regulation of cell growth
positive regulation of cell migration
microtubule anchoring
negative regulation of protein tyrosine kinase activity
protein kinase binding
protein tyrosine kinase inhibitor activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0