This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06396155_g1
See other IDI2 GE Assays ›
Gene Symbol:
Idi2 LOC103694120
Gene Name:
isopentenyl-diphosphate delta isomerase 2
isopentenyl-diphosphate delta-isomerase 2-like
Gene Aliases:
RGD1559783, RGD1562352
-
Chromosome Location:
Chr.17: 63107152 - 63107469 on Build Rnor_6.0
Chromosome Location:
Chr.17: 57814239 - 57816262 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn06396155_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idi2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001192008.1 NP_001178937.1 365 79
Gene Symbol: LOC103694120
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008771843.1 XP_008770065.1 2 - 3 230 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

isoprenoid biosynthetic process
isopentenyl-diphosphate delta-isomerase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0