This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06392121_s1
See other CHRM3 GE Assays ›
Gene Symbol:
Chrm3
Gene Name:
cholinergic receptor, muscarinic 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 63990599 - 64463222 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06392121_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chrm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012527.2 NP_036659.2 1 198 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_012527.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Chrm3

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

regulation of vascular smooth muscle contraction
smooth muscle contraction
G-protein coupled acetylcholine receptor signaling pathway
digestion
regulation of vasoconstriction
positive regulation of smooth muscle contraction
saliva secretion
protein binding
drug binding
G-protein coupled acetylcholine receptor activity
acetylcholine binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0