This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06357779_g1
See other CALM2 GE Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06357779_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Calm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC058485.1 - 240 6921326 62
FQ211494.1 - 240 62
FQ211970.1 - 236 62
FQ212579.1 - 236 62
FQ212640.1 - 236 62
FQ212691.1 - 236 62
FQ212789.1 - 240 62
FQ212869.1 - 240 62
FQ213028.1 - 236 62
FQ213029.1 - 234 62
FQ213091.1 - 236 62
FQ213115.1 - 242 62
FQ213200.1 - 239 62
FQ213216.1 - 236 62
FQ213310.1 - 129 62
FQ213366.1 - 235 62
FQ213496.1 - 239 62
FQ213520.1 - 240 62
FQ213526.1 - 236 62
FQ213581.1 - 236 62
FQ213598.1 - 236 62
FQ213637.1 - 236 62
FQ213703.1 - 236 62
FQ213760.1 - 236 62
FQ213762.1 - 241 62
FQ213913.1 - 236 62
FQ213941.1 - 240 62
FQ213981.1 - 237 62
FQ213985.1 - 130 62
FQ214059.1 - 240 62
FQ214181.1 - 240 62
FQ214262.1 - 240 62
FQ214491.1 - 236 62
FQ214500.1 - 236 62
FQ214519.1 - 236 62
FQ214525.1 - 236 62
FQ214540.1 - 240 62
FQ215159.1 - 240 62
FQ215317.1 - 236 62
FQ215600.1 - 240 62
FQ216397.1 - 236 62
FQ216471.1 - 237 62
FQ218091.1 - 236 62
FQ220250.1 - 236 62
FQ220325.1 - 236 62
FQ220698.1 - 240 62
FQ220779.1 - 236 62
FQ222184.1 - 239 62
FQ223357.1 - 240 62
FQ225272.1 - 236 62
FQ226994.1 - 240 62
FQ229228.1 - 236 62
FQ229580.1 - 240 62
FQ231022.1 - 240 62
FQ231702.1 - 240 62
FQ233337.1 - 239 62
FQ234377.1 - 240 62
FQ235266.1 - 237 62
M17069.1 - 238 62
M19312.1 - 219 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus

Panther Classification:

Molecular Function -

calmodulin-related

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0