This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06307588_g1
See other LOC100911403 GE Assays ›
Gene Symbol:
LOC100911403 Ms4a4a
Gene Name:
membrane-spanning 4-domains subfamily A member 4A-like
membrane-spanning 4-domains, subfamily A, member 4A
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 227217927 - 227232337 on Build Rnor_6.0
Chromosome Location:
Chr.1: 227575454 - 227606958 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn06307588_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100911403
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006231085.2 XP_006231147.1 6 803 103
XM_008760268.1 XP_008758490.1 5 684 103
Gene Symbol: Ms4a4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008760273.1 XP_008758495.1 8 3500 103
XM_008760275.1 XP_008758497.1 6 4986 103
XM_342027.5 XP_342028.3 7 5115 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0