This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06304206_s1
See other SUPT3H GE Assays ›
Gene Symbol:
Supt3h
Gene Name:
suppressor of Ty 3 homolog (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.9: 18249565 - 18604790 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06304206_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Supt3h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006244587.2 XP_006244649.2 12 11961 67
XM_008766906.1 XP_008765128.1 12 11980 67
XM_008766907.1 XP_008765129.1 12 11453 67
XM_008766908.1 XP_008765130.1 12 11349 67
XM_008766909.1 XP_008765131.1 12 11310 67
XM_008766910.1 XP_008765132.1 11 11223 67
XM_008766911.1 XP_008765133.1 11 11208 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
biological_process
histone acetylation
histone deubiquitination
histone H3 acetylation
molecular_function
DNA binding
transcription cofactor activity
transcription coactivator activity
histone acetyltransferase activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header