This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06265641_g1
See other CRP GE Assays ›
Gene Symbol:
Crp LOC100233206
Gene Name:
C-reactive protein, pentraxin-related
LINE-1 retrotransposable element ORF2 protein
Gene Aliases:
Aa1249, Ab1-341, Ab2-196, Ac1-114, Ac1262, Ac2-069, Ba2-693
-
Chromosome Location:
Chr.13: 91080448 - 91081358 on Build Rnor_6.0
Chromosome Location:
Chr.3: 145301953 - 145312333 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn06265641_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY310153.1 - 617 106
AY318961.1 - 617 106
AY321318.1 - 614 106
AY321324.1 - 617 106
AY325196.1 - 617 106
AY325218.1 - 617 106
AY325232.1 - 617 106
Gene Symbol: LOC100233206
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY539901.1 - 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus

Panther Classification:

Molecular Function -

reverse transcriptase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0