This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06254576_g1
See other LOC100362110 GE Assays ›
Gene Symbol:
LOC100362110 Crp LOC501503 Lrrgt00014 Serinc1
Gene Name:
LRRGT00155-like
C-reactive protein, pentraxin-related
nucleic acid binding protein 5
uncharacterized LOC361915
serine incorporator 1
Gene Aliases:
LRRGT00155
Aa1249, Ab1-341, Ab2-196, Ac1-114, Ac1262, Ac2-069, Ba2-693
-
-
LRRGT00191, Tde1l, Tde2
Chromosome Location:
Chr.13: 41802547 - 41836153 on Build Rnor_6.0
Chromosome Location:
Chr.13: 91080448 - 91081358 on Build Rnor_6.0
Chromosome Location:
-
Chromosome Location:
-
Chromosome Location:
Chr.20: 38966016 - 38985056 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
-
-
Assay ID Rn06254576_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100362110
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA S80119.1 - 755 106
Gene Symbol: Crp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY310153.1 - 781 106
AY318961.1 - 781 106
AY321318.1 - 778 106
AY321324.1 - 781 106
AY325196.1 - 781 106
AY325218.1 - 781 106
AY325232.1 - 781 106
Gene Symbol: LOC501503
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA DQ100477.1 - 2 4394 106
Gene Symbol: Lrrgt00014
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA U83119.1 - 1 2697 106
Gene Symbol: Serinc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY539942.1 - 1992 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

reverse transcriptase viral or transposable element protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0