This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06247052_g1
See other RGD1565987 GE Assays ›
Gene Symbol:
RGD1565987 Fbxl18 LOC100909844
Gene Name:
similar to F-box and leucine-rich repeat protein 18
F-box and leucine-rich repeat protein 18
F-box/LRR-repeat protein 18-like
Gene Aliases:
-
-
-
Chromosome Location:
Chr.5: 98569723 - 98571614 on Build Rnor_6.0
Chromosome Location:
Chr.12: 13728780 - 13751781 on Build Rnor_6.0
Chromosome Location:
Chr.5: 98546574 - 98548714 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
Assay ID Rn06247052_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGD1565987
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006238333.2 XP_006238395.1 1 - 2 901 70
Gene Symbol: Fbxl18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006248907.2 XP_006248969.1 3 - 4 1874 70
Gene Symbol: LOC100909844
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003749966.3 XP_003750014.2 1 - 2 1829 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0