This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06198078_m1
See other CRAMP1L GE Assays ›
Gene Symbol:
Cramp1l
Gene Name:
Crm, cramped-like (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.10: 14324799 - 14373411 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn06198078_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cramp1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006246051.2 XP_006246113.1 6 - 7 1463 91
XM_006246052.2 XP_006246114.1 6 - 7 1127 91
XM_006246053.2 XP_006246115.1 6 - 7 913 91
XM_008767606.1 XP_008765828.1 6 - 7 1463 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Process(es)

chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0