This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06179038_g1
See other RGD1560917 GE Assays ›
Gene Symbol:
RGD1560917 Mrpl41
Gene Name:
similar to mitochondrial ribosomal protein L41
mitochondrial ribosomal protein L41
Gene Aliases:
-
-
Chromosome Location:
Chr.8: 91820577 - 91821318 on Build Rnor_6.0
Chromosome Location:
Chr.3: 2181044 - 2181962 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn06179038_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGD1560917
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003750552.3 XP_003750600.1 1 318 103
Gene Symbol: Mrpl41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013426.1 NP_001013444.1 2 282 103
GenBank mRNA BC091293.1 - 2 282 7307323 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RGD1560917
Click here to find siRNAs for Gene Mrpl41

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
apoptotic process
structural constituent of ribosome
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0