This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn06156843_g1
See other LOC100363472 GE Assays ›
Gene Symbol:
LOC100363472 Nrbf2 RGD1560263
Gene Name:
nuclear receptor-binding factor 2-like
nuclear receptor binding factor 2
similar to nuclear receptor binding factor 2
Gene Aliases:
-
-
-
Chromosome Location:
Chr.14: 10624057 - 10625903 on Build Rnor_6.0
Chromosome Location:
Chr.20: 22728147 - 22746477 on Build Rnor_6.0
Chromosome Location:
Chr.2: 195855521 - 195857095 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
Assay ID Rn06156843_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100363472
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002728074.4 XP_002728120.1 1 509 75
XM_008770073.1 XP_008768295.1 2 566 75
Gene Symbol: Nrbf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022186.1 NP_071522.1 4 561 75
XM_008772913.1 XP_008771135.1 3 537 75
GenBank mRNA AB024930.1 - 4 561 75
Gene Symbol: RGD1560263
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008761340.1 XP_008759562.1 2 473 75
XM_574989.5 XP_574989.1 1 456 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LOC100363472
Click here to find siRNAs for Gene Nrbf2
Click here to find siRNAs for Gene RGD1560263

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s)

transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of ligand-dependent nuclear receptor transcription coactivator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0