This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04932594_m1
See other LOC100909680 GE Assays ›
Gene Symbol:
LOC100909680
Gene Name:
uncharacterized LOC100909680
Gene Aliases:
Lrrgt00201
Chromosome Location:
Chr.6: 101864565 - 101873464 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04932594_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100909680
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY539952.1 - 2 - 3 166 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream gephyrin Gphn Geph -5.8kb Anti-sense Chr. 6 - 101327874 - 101859169 on NCBI Build Rnor_6.0
Downstream membrane protein,
palmitoylated 5 (MAGUK p55 subfamily member 5)
Mpp5 55.6kb Anti-sense Chr. 6 - 101923670 - 102029705 on NCBI Build Rnor_6.0

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0