This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04418762_m1
See other CDKN2B GE Assays ›
Gene Symbol:
Cdkn2b
Gene Name:
cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4)
Gene Aliases:
p15
Chromosome Location:
Chr.5: 107848177 - 107857323 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04418762_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdkn2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130812.3 NP_570825.1 1 - 2 316 65
XM_006238380.2 XP_006238442.1 1 - 2 248 65
XM_006238381.1 XP_006238443.1 1 - 2 248 65
GenBank mRNA AF474978.1 - 1 - 2 230 65
AF474979.1 - 1 - 2 232 65
BC086995.1 - 1 - 2 251 7134283 65
CK603883.1 - 1 - 2 254 65
S79760.1 - 1 - 2 291 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdkn2b

Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
G2/M transition of mitotic cell cycle
liver development
cell cycle arrest
mitotic cell cycle checkpoint
aging
negative regulation of cell proliferation
response to organic cyclic compound
megakaryocyte differentiation
positive regulation of transforming growth factor beta receptor signaling pathway
positive regulation of epithelial cell differentiation
cellular response to extracellular stimulus
cellular response to nutrient
response to cytokine
negative regulation of phosphorylation
spleen development
negative regulation of epithelial cell proliferation
negative regulation of protein serine/threonine kinase activity
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0