This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04341320_m1
See other IGFBP6 GE Assays ›
Gene Symbol:
Igfbp6
Gene Name:
insulin-like growth factor binding protein 6
Gene Aliases:
IGF-BP6, RBP6A
Chromosome Location:
Chr.7: 143749385 - 143754018 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04341320_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igfbp6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013104.3 NP_037236.1 1 - 2 307 100
GenBank mRNA BC099742.1 - 1 - 2 319 7937967 100
FQ220221.1 - 1 - 2 377 100
FQ220764.1 - 1 - 2 380 100
FQ230155.1 - 1 - 2 383 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700001891.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igfbp6

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
biological_process
regulation of insulin-like growth factor receptor signaling pathway
insulin-like growth factor I binding
insulin-like growth factor II binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700014731.rn.1 TC0700001891.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0