This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244865_m1
See other SFTPD GE Assays ›
Gene Symbol:
Sftpd
Gene Name:
surfactant protein D
Gene Aliases:
SP-D, SPD
Chromosome Location:
Chr.16: 18753456 - 18766100 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04244865_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sftpd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012878.2 NP_037010.1 2 - 3 333 82
XM_008771065.1 XP_008769287.1 2 - 3 336 82
GenBank mRNA BC070507.1 - 3 - 4 342 7098194 82
M81231.1 - 3 - 4 340 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sftpd

Panther Classification:

Molecular Function -

surfactant

Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
opsonization
response to corticosteroid
negative regulation of interleukin-2 production
negative regulation of T cell proliferation
surfactant homeostasis
innate immune response
lung alveolus development
negative regulation of phagocytosis
positive regulation of phagocytosis
regulation of liquid surface tension
response to glucocorticoid
response to hyperoxia
response to growth factor
lipopolysaccharide binding
carbohydrate binding
identical protein binding
monosaccharide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0