This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244707_m1
See other S100Z GE Assays ›
Gene Symbol:
S100z
Gene Name:
S100 calcium binding protein Z
Gene Aliases:
RGD1562902
Chromosome Location:
Chr.2: 25188389 - 25192552 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04244707_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100z
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191701.1 NP_001178630.1 1 - 2 136 76
XM_008760662.1 XP_008758884.1 3 - 4 928 76
XM_008760663.1 XP_008758885.1 3 - 4 1029 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200002646.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

calcium ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P508653 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508653 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508653 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P508653 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508653 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200018508.rn.1 TC0200002646.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200018509.rn.1 TC0200002646.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1369418 - -
RatRef-12 Expression BeadChip ILMN_60877 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0