This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244643_s1
See other TMEM185B GE Assays ›
Gene Symbol:
Tmem185b
Gene Name:
transmembrane protein 185B
Gene Aliases:
RGD1309602
Chromosome Location:
Chr.13: 35554608 - 35557952 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04244643_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem185b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191668.3 NP_001178597.1 1 1285 79

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P180240 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P180240 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P180240 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P180240 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P180240 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10763549 - -
Affymetrix Rat Gene 1.0 ST Array 10763548 - -
Rat Genome 230 2.0 Array 1390392_PM_at - -
Rat Genome 230 2.0 Array 1390392_at - -
Affymetrix HT Rat Focus Array 1390392_at - -
Affymetrix HT Rat Focus Array 1390392_PM_at - -
Affymetrix Rat Clariom S Assay PSR1300000717.rn.1 TC1300000137.rn.1 RTA TC
Rat Expression Set 230 1390392_PM_at - -
Rat Expression Set 230 1390392_at - -
Affymetrix Rat Gene 1.0 ST Array 10763547 - -
Rat Genome U34 Set rc_AI145722_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21428131 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1352654 - -
RatRef-12 Expression BeadChip ILMN_53153 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0