This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04244078_s1
See other MIR7A-2 GE Assays ›
Gene Symbol:
Mir7a-2
Gene Name:
microRNA mir-7a-2
Gene Aliases:
rno-mir-7a-2
Chromosome Location:
Chr.1: 140576396 - 140576490 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04244078_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir7a-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031809.1 1 49 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream de-etiolated homolog 1 (Arabidopsis) Det1 RGD1309793 -40.4kb Anti-sense Chr. 1 - 140519755 - 140535970 on NCBI Build Rnor_6.0
Downstream apoptosis enhancing nuclease Aen Isg20l1,
RGD1305051
7.9kb Sense Chr. 1 - 140584343 - 140593749 on NCBI Build Rnor_6.0

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

posttranscriptional regulation of gene expression
gene silencing by miRNA
SMAD protein signal transduction
intestinal epithelial cell development
cellular response to lipopolysaccharide
cellular response to inorganic substance

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100014257.rn.1 TC0100001820.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0