This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04224388_u1
See other SLC39A4L GE Assays ›
Gene Symbol:
Slc39a4l
Gene Name:
solute carrier family 39 (zinc transporter), member 4-like
Gene Aliases:
Slc39a10, Zip10
Chromosome Location:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04224388_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc39a4l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101021.1 NP_001094491.1 1254 89
GenBank mRNA DQ256461.1 - 1254 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus

Panther Classification:

Molecular Function -

secondary carrier transporter

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009823 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009823 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009823 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009823 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10766775 - -
Affymetrix Rat Gene 1.1 ST Array 10766774 - -
Affymetrix Rat Gene 1.0 ST Array 10766774 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0