This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn04222691_gH
See other LILRA5 GE Assays ›
Gene Symbol:
Lilra5
Gene Name:
leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5
Gene Aliases:
Lilrc1
Chromosome Location:
Chr.1: 73655775 - 73660941 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn04222691_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lilra5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076793.1 NP_001070261.1 1 - 2 39 71
GenBank mRNA AY998481.1 - 1 - 2 39 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY998481.1, NM_001076793.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100037937.rn.1 TC0100004811.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100037938.rn.1 TC0100004811.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0