Assay Details
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031832.1 | NP_114020.1 | 3 - 4 | 410 | 65 | |
GenBank mRNA | BC089054.1 | - | 3 - 4 | 398 | 7312891 | 65 |
FQ214799.1 | - | 3 - 4 | 692 | 65 | ||
FQ218337.1 | - | 3 - 4 | 410 | 65 | ||
FQ219929.1 | - | 3 - 4 | 410 | 65 | ||
FQ219987.1 | - | 3 - 4 | 410 | 65 | ||
FQ220157.1 | - | 3 - 4 | 410 | 65 | ||
FQ220174.1 | - | 3 - 4 | 409 | 65 | ||
FQ220215.1 | - | 3 - 4 | 412 | 65 | ||
FQ220224.1 | - | 3 - 4 | 404 | 65 | ||
FQ220313.1 | - | 3 - 4 | 410 | 65 | ||
FQ220316.1 | - | 4 - 5 | 640 | 65 | ||
FQ220348.1 | - | 3 - 4 | 410 | 65 | ||
FQ220361.1 | - | 3 - 4 | 569 | 65 | ||
FQ220369.1 | - | 3 - 4 | 410 | 65 | ||
FQ220439.1 | - | 3 - 4 | 409 | 65 | ||
FQ220450.1 | - | 3 - 4 | 410 | 65 | ||
FQ220455.1 | - | 4 - 5 | 637 | 65 | ||
FQ220487.1 | - | 4 - 5 | 610 | 65 | ||
FQ220505.1 | - | 3 - 4 | 410 | 65 | ||
FQ220541.1 | - | 3 - 4 | 410 | 65 | ||
FQ220563.1 | - | 3 - 4 | 410 | 65 | ||
FQ220590.1 | - | 3 - 4 | 412 | 65 | ||
FQ220652.1 | - | 3 - 4 | 410 | 65 | ||
FQ220701.1 | - | 3 - 4 | 409 | 65 | ||
FQ220837.1 | - | 3 - 4 | 409 | 65 | ||
FQ220917.1 | - | 3 - 4 | 410 | 65 | ||
FQ220938.1 | - | 3 - 4 | 410 | 65 | ||
FQ220946.1 | - | 3 - 4 | 410 | 65 | ||
FQ220952.1 | - | 3 - 4 | 410 | 65 | ||
FQ221377.1 | - | 3 - 4 | 410 | 65 | ||
FQ223467.1 | - | 3 - 4 | 412 | 65 | ||
FQ223531.1 | - | 3 - 4 | 410 | 65 | ||
FQ229211.1 | - | 3 - 4 | 414 | 65 | ||
FQ229249.1 | - | 3 - 4 | 410 | 65 | ||
FQ229259.1 | - | 3 - 4 | 410 | 65 | ||
FQ229266.1 | - | 3 - 4 | 410 | 65 | ||
FQ229277.1 | - | 3 - 4 | 407 | 65 | ||
FQ229326.1 | - | 3 - 4 | 410 | 65 | ||
FQ229348.1 | - | 3 - 4 | 410 | 65 | ||
FQ229362.1 | - | 3 - 4 | 410 | 65 | ||
FQ229490.1 | - | 3 - 4 | 410 | 65 | ||
FQ229494.1 | - | 3 - 4 | 410 | 65 | ||
FQ229511.1 | - | 3 - 4 | 410 | 65 | ||
FQ229560.1 | - | 3 - 4 | 412 | 65 | ||
FQ229562.1 | - | 3 - 4 | 412 | 65 | ||
FQ229564.1 | - | 3 - 4 | 412 | 65 | ||
FQ229577.1 | - | 3 - 4 | 410 | 65 | ||
FQ229661.1 | - | 3 - 4 | 410 | 65 | ||
FQ229821.1 | - | 3 - 4 | 410 | 65 | ||
FQ229967.1 | - | 3 - 4 | 412 | 65 | ||
FQ229990.1 | - | 3 - 4 | 412 | 65 | ||
FQ229991.1 | - | 3 - 4 | 412 | 65 | ||
FQ229995.1 | - | 3 - 4 | 410 | 65 | ||
FQ230027.1 | - | 3 - 4 | 410 | 65 | ||
FQ230050.1 | - | 3 - 4 | 413 | 65 | ||
FQ230072.1 | - | 3 - 4 | 410 | 65 | ||
FQ230121.1 | - | 3 - 4 | 417 | 65 | ||
FQ230149.1 | - | 3 - 4 | 412 | 65 | ||
J02962.1 | - | 3 - 4 | 410 | 65 | ||
KF639951.1 | - | 2 - 3 | 370 | 65 | ||
KF639952.1 | - | 2 - 3 | 370 | 65 | ||
KF639953.1 | - | 2 - 3 | 370 | 65 | ||
KF639957.1 | - | 3 - 4 | 349 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
