This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03652673_s1
See other SFTA2 GE Assays ›
Gene Symbol:
Sfta2
Gene Name:
surfactant associated 2
Gene Aliases:
E030032D13Rik
Chromosome Location:
Chr.20: 3604918 - 3605643 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03652673_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sfta2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166020.1 NP_001159492.1 3 239 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BF521842.1 - 241 Chr. - - [-] on Build 75
GenBankmRNA AI136523.1 - Chr. - - [-] on Build 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream valyl-tRNA synthetase 2,
mitochondrial
Vars2 Vars2l,
Varsl
-5.4kb Anti-sense Chr. 20 - 3588460 - 3599514 on NCBI Build Rnor_6.0
Downstream diffuse panbronchiolitis critical region 1 Dpcr1 6.9kb Anti-sense Chr. 20 - 3611965 - 3619884 on NCBI Build Rnor_6.0

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

surfactant

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0