This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03651254_s1
See other MIR199A2 GE Assays ›
Gene Symbol:
Mir199a2
Gene Name:
microRNA 199a-2
Gene Aliases:
Mir199a, rno-mir-199a
Chromosome Location:
Chr.13: 80125487 - 80125596 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03651254_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir199a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031914.1 1 57 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA FQ140697.1 - 207 Chr. - - [-] on Build 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream similar to hypothetical protein RGD1309106 -226.0kb Anti-sense Chr. 13 - 79856479 - 79899479 on NCBI Build Rnor_6.0
Downstream microRNA mir-214 Mir214 rno-mir-214 5.3kb Sense Chr. 13 - 80130905 - 80131014 on NCBI Build Rnor_6.0

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
negative regulation of fatty acid oxidation
response to oxygen levels
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to inorganic substance
cellular response to hypoxia
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300004339.rn.1 TC1300000622.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0