This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03416150_m1
See other DACT2 GE Assays ›
Gene Symbol:
Dact2
Gene Name:
dishevelled-binding antagonist of beta-catenin 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 56159435 - 56169427 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03416150_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dact2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107464.2 NP_001100934.2 1 - 2 239 140
GenBank mRNA CB788616.1 - 1 - 2 307 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100004554.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
epithelial cell morphogenesis
negative regulation of cell adhesion
regulation of Wnt signaling pathway
skin development
inner medullary collecting duct development
negative regulation of nodal signaling pathway
protein kinase C binding
beta-catenin binding
transcription factor binding
protein kinase A binding
delta-catenin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100036041.rn.1 TC0100004554.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100036050.rn.1 TC0100004554.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0