This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02117596_s1
See other CREBL2 GE Assays ›
Gene Symbol:
Crebl2
Gene Name:
cAMP responsive element binding protein-like 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 168599239 - 168625239 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02117596_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crebl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015027.1 NP_001015027.1 5 84
GenBank mRNA BC091323.1 - 5 7313598 84

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crebl2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cell differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of glucose import
positive regulation of lipid biosynthetic process
protein stabilization
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400013910.rn.1 TC0400001923.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20763458 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0