This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01771561_m1
See other SMC4 GE Assays ›
Gene Symbol:
Smc4
Gene Name:
structural maintenance of chromosomes 4
Gene Aliases:
Smc4l1
Chromosome Location:
Chr.2: 165601007 - 165629415 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01771561_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037185.2 NP_001032262.2 5 - 6 995 84
GenBank mRNA BC107944.1 - 5 - 6 769 84
FQ226690.1 - 3 - 4 399 84
FQ227460.1 - 2 - 3 258 84
FQ230642.1 - 2 - 3 316 84
FQ233743.1 - 3 - 4 399 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200001360.rn.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitotic chromosome condensation
meiotic chromosome condensation
meiotic chromosome segregation
kinetochore organization
ATP binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200008474.rn.1 TC0200001360.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0