This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01771103_m1
See other IER3IP1 GE Assays ›
Gene Symbol:
Ier3ip1
Gene Name:
immediate early response 3 interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.4: 132140802 - 132151787 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01771103_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ier3ip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001047956.1 NP_001041421.1 2 - 3 118 88
XM_008763163.1 XP_008761385.1 1 - 2 113 88
GenBank mRNA AY325263.1 - 2 - 3 118 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400001484.rn.1

Set Membership:

Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s)

regulation of fibroblast apoptotic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400010430.rn.1 TC0400001484.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0400010433.rn.1 TC0400001484.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0