This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01767898_m1
See other CTDSP2 GE Assays ›
Gene Symbol:
Ctdsp2
Gene Name:
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 70283689 - 70291795 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01767898_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctdsp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006241502.2 XP_006241564.1 3 - 4 301 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006241502.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
protein dephosphorylation
negative regulation of G1/S transition of mitotic cell cycle
CTD phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0