This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01765151_m1
See other CAMK1D GE Assays ›
Gene Symbol:
Camk1d
Gene Name:
calcium/calmodulin-dependent protein kinase ID
Gene Aliases:
RGD1560691
Chromosome Location:
Chr.17: 76532611 - 76938956 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01765151_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camk1d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107365.1 NP_001100835.1 10 - 11 1075 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700000992.rn.1

Set Membership:

Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
positive regulation of neuron projection development
positive regulation of CREB transcription factor activity
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of phagocytosis
regulation of dendrite development
positive regulation of respiratory burst
regulation of granulocyte chemotaxis
positive regulation of neutrophil chemotaxis
protein serine/threonine kinase activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700006836.rn.1 TC1700000992.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0