This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01763461_m1
See other GPR128 GE Assays ›
Gene Symbol:
Gpr128 LOC100911145
Gene Name:
G protein-coupled receptor 128
probable G-protein coupled receptor 128-like
Gene Aliases:
-
-
Chromosome Location:
Chr.11: 46793777 - 46856291 on Build Rnor_6.0
Chromosome Location:
Chr.11: 46107178 - 46172303 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01763461_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr128
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107098.1 NP_001100568.1 1478 77
Gene Symbol: LOC100911145
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003751038.3 XP_003751086.1 10 - 11 1306 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100000471.rn.1

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1100002341.rn.1 TC1100000471.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0