This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01763407_g1
See other GPR15 GE Assays ›
Gene Symbol:
Gpr15
Gene Name:
G protein-coupled receptor 15
Gene Aliases:
-
Chromosome Location:
Chr.11: 44001579 - 44003664 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01763407_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105890.1 NP_001099360.1 1 - 2 1439 192

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100000443.rn.1

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
T cell migration
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P462158 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 572752 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 466671 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P462158 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P462158 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P462158 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P462158 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10750660 - -
Affymetrix Rat Gene 1.1 ST Array 10750659 - -
Affymetrix Rat Clariom S Assay PSR1100002101.rn.1 TC1100000443.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10750659 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22294582 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1352981 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0