This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01763334_m1
See other HEG1 GE Assays ›
Gene Symbol:
Heg1
Gene Name:
heart development protein with EGF-like domains 1
Gene Aliases:
RGD1564641
Chromosome Location:
Chr.11: 70234248 - 70322962 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01763334_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Heg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006248443.2 XP_006248505.1 15 - 16 3955 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
in utero embryonic development
endothelial cell development
endothelial cell morphogenesis
vasculature development
lymph vessel development
lymph circulation
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular septum development
cell-cell junction assembly
heart development
post-embryonic development
lung development
multicellular organism growth
cell-cell junction organization
venous blood vessel morphogenesis
regulation of body fluid levels
cardiac muscle tissue growth
pericardium development
positive regulation of fibroblast growth factor production
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0