This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01761850_m1
See other GPD1 GE Assays ›
Gene Symbol:
Gpd1
Gene Name:
glycerol-3-phosphate dehydrogenase 1 (soluble)
Gene Aliases:
GPDH, Gpd3
Chromosome Location:
Chr.7: 141369490 - 141377931 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01761850_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022215.2 NP_071551.2 2 - 3 234 81
XM_006257376.1 XP_006257438.1 3 - 4 436 81
GenBank mRNA AB002558.1 - 2 - 3 222 81
BC088396.1 - 2 - 3 234 7108237 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700001823.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpd1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycerol-3-phosphate metabolic process
gluconeogenesis
NADH oxidation
glycerophosphate shuttle
NADH metabolic process
positive regulation of glycolytic process
glycerol-3-phosphate catabolic process
glycerolipid metabolic process
oxidation-reduction process
cellular response to cAMP
cellular response to tumor necrosis factor
glycerol-3-phosphate dehydrogenase [NAD+] activity
glycerol-3-phosphate dehydrogenase activity
protein homodimerization activity
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700014093.rn.1 TC0700001823.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0